Assay Details
Target Gene Details
Entrez Gene ID: | 84818 |
Gene Name: | interleukin 17 receptor C |
Gene Aliases: |
CANDF9, IL17-RL, IL17RL |
Location: |
Chr.3:9917074-9933627 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
| nsv589648 | Chr.3:9892921 - 10042251 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map