Assay Details
Target Gene Details
Entrez Gene ID: | 51147 |
Gene Name: | inhibitor of growth family member 4 |
Gene Aliases: |
my036, p29ING4 |
Location: |
Chr.12:6650538-6663143 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509452 | Chr.12:6566935 - 6653627 on Build GRCh38 | Insertion |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map