Assay Details
Target Gene Details
Entrez Gene ID: | 11321 |
Gene Name: | GPN-loop GTPase 1 |
Gene Aliases: |
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1 |
Location: |
Chr.2:27628648-27650846 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPN1 | NM_001145047.1 | 10 | 644 | NP_001138519.1 |
| NM_001145048.1 | 9 | 576 | NP_001138520.1 | |
| NM_001145049.1 | 7 | 469 | NP_001138521.1 | |
| NM_007266.3 | 9 | 683 | NP_009197.2 | |
| NR_026735.1 | 7 | 576 | ||
| AB044661.1 | 9 | 644 | BAB17612.1 | |
| AJ010842.1 | 9 | 573 | CAA09376.1 | |
| AK298827.1 | 9 | 977 | ||
| AK298866.1 | 9 | 683 | ||
| AK302131.1 | 7 | 542 | ||
| AK310340.1 | 10 | 634 | ||
| AK311423.1 | 7 | 599 | ||
| AK312992.1 | 9 | 655 | ||
| AK316404.1 | 7 | 469 | ||
| BC007451.2 | 9 | 627 | AAH07451.1 |
More Information
Set Membership: |
|

Genomic Map