Assay Details
Target Gene Details
Entrez Gene ID: | 402055 |
Gene Name: | SRR1 domain containing |
Gene Aliases: |
HC/HCC, SRR1L |
Location: |
Chr.22:26483867-26494620 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRD | NM_001013694.2 | 7 | 1239 | NP_001013716.2 |
| XM_011530178.2 | 7 | 1302 | XP_011528480.1 | |
| XM_017028799.1 | 6 | 1096 | XP_016884288.1 | |
| AK293916.1 | 8 | 1219 | ||
| BC066962.1 | 8 | 1222 | AAH66962.1 | |
| BQ189353.1 | 1 | 376 |
More Information
Additional Information:
For this assay, SNP(s) [rs115358886] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map