Assay Details
Target Gene Details
Entrez Gene ID: | 255762 |
Gene Name: | PDZ domain containing 9 |
Gene Aliases: |
C16orf65 |
Location: |
Chr.16:21957540-22001137 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZD9 | XM_011545785.2 | XP_011544087.1 | ||
| XM_017023109.1 | XP_016878598.1 | |||
| XM_017023110.1 | XP_016878599.1 |
Target Gene Details
Entrez Gene ID: | 7385 |
Gene Name: | ubiquinol-cytochrome c reductase core protein II |
Gene Aliases: |
MC3DN5, QCR2, UQCR2 |
Location: |
Chr.16:21953064-21983660 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCRC2 | NM_003366.3 | NP_003357.2 | ||
| AK094006.1 | ||||
| AK307535.1 | ||||
| BC000484.2 | AAH00484.1 | |||
| BC003136.1 | AAH03136.1 | |||
| J04973.1 | AAA35710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34235 | Chr.16:21276264 - 21986230 on Build GRCh38 | Gain |
|
| nsv1048054 | Chr.16:21952632 - 21994369 on Build GRCh38 | Gain |
|
| nsv428322 | Chr.16:21489241 - 22017793 on Build GRCh38 | Gain |
|
| esv3638193 | Chr.16:21934532 - 22029031 on Build GRCh38 | Gain |
|
| esv34544 | Chr.16:21522978 - 21986230 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map