Assay Details
Target Gene Details
Entrez Gene ID: | 9344 |
Gene Name: | TAO kinase 2 |
Gene Aliases: |
MAP3K17, PSK, PSK1, PSK1-BETA, TAO1, TAO2 |
Location: |
Chr.16:29973641-29992261 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAOK2 | NM_001252043.1 | 12 | 1902 | NP_001238972.1 |
| NM_004783.3 | 12 | 1902 | NP_004774.1 | |
| NM_016151.3 | 12 | 1902 | NP_057235.2 | |
| XM_011545982.1 | 12 | 2149 | XP_011544284.1 | |
| XM_011545983.2 | 12 | 2146 | XP_011544285.1 | |
| XM_011545984.1 | 12 | 2149 | XP_011544286.1 | |
| XM_011545985.1 | 12 | 2146 | XP_011544287.1 | |
| XM_011545986.2 | 12 | 2149 | XP_011544288.1 | |
| AB020688.1 | 12 | 1488 | ||
| AF061943.1 | 12 | 1843 | AAD45616.1 | |
| AF263313.1 | 11 | 1085 | AAG38503.1 | |
| AK291473.1 | 12 | 1419 | ||
| AK304446.1 | 4 | 408 | ||
| AL137701.1 | 4 | 1739 | CAB70882.1 | |
| BC015204.1 | 12 | 1868 | ||
| BC031825.2 | 12 | 1160 | AAH31825.1 | |
| BC051798.1 | 12 | 1659 | AAH51798.1 | |
| BC136653.1 | 12 | 1587 | ||
| BC136655.1 | 12 | 1587 | ||
| BC142663.1 | 12 | 1242 | ||
| BC144344.1 | 12 | 1587 | ||
| BC151221.1 | 12 | 1242 | ||
| BC152413.1 | 12 | 1488 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map