Assay Details
Target Gene Details
Entrez Gene ID: | 53335 |
Gene Name: | B-cell CLL/lymphoma 11A |
Gene Aliases: |
BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856 |
Location: |
Chr.2:60451167-60553498 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL11A | NM_018014.3 | 5 | 2849 | NP_060484.2 |
| NM_138559.1 | 5 | 1249 | NP_612569.1 | |
| XM_011532909.1 | 5 | 3129 | XP_011531211.1 | |
| XM_011532910.1 | 5 | 3001 | XP_011531212.1 | |
| XM_011532912.1 | 4 | 2899 | XP_011531214.1 | |
| AB058712.1 | 5 | 2749 | ||
| AF080216.1 | 7 | 2929 | AAG49025.1 | |
| AJ404612.1 | 5 | 2849 | CAC17724.1 | |
| AJ404613.1 | 5 | 1249 | CAC17725.1 | |
| AK055251.1 | 5 | 2867 | ||
| BC021098.1 | 5 | 2730 | AAH21098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834204 | Chr.2:60303743 - 60474590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map