Assay Details
Target Gene Details
Entrez Gene ID: | 91782 |
Gene Name: | charged multivesicular body protein 7 |
Gene Aliases: |
- |
Location: |
Chr.8:23243637-23262000 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP7 | NM_001317899.1 | NP_001304828.1 | ||
| NM_152272.4 | NP_689485.1 | |||
| XM_006716415.2 | XP_006716478.1 | |||
| XM_017013961.1 | XP_016869450.1 | |||
| XM_017013962.1 | XP_016869451.1 | |||
| XM_017013963.1 | XP_016869452.1 | |||
| XM_017013964.1 | XP_016869453.1 | |||
| XM_017013965.1 | XP_016869454.1 | |||
| AK021776.1 | ||||
| AK092500.1 | ||||
| AK097202.1 | ||||
| AK296592.1 | ||||
| AK315664.1 | ||||
| AL833843.1 | CAD38703.2 | |||
| BC004344.1 | AAH04344.1 | |||
| BC019110.1 | AAH19110.1 | |||
| BC042050.1 | AAH42050.1 | |||
| DA773200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027718 | Chr.8:22805950 - 23258334 on Build GRCh38 | Gain |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
| nsv1017570 | Chr.8:22969562 - 23544783 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map