Assay Details
Target Gene Details
Entrez Gene ID: | 151393 |
Gene Name: | regulator of microtubule dynamics 2 |
Gene Aliases: |
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4 |
Location: |
Chr.2:37920789-38068032 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN2 | NM_001170791.2 | NP_001164262.1 | ||
| NM_001170792.2 | NP_001164263.1 | |||
| NM_001170793.2 | 1 | 68 | NP_001164264.1 | |
| NM_001322211.1 | NP_001309140.1 | |||
| NM_001322212.1 | NP_001309141.1 | |||
| NM_144713.4 | 1 | 68 | NP_653314.3 | |
| XM_011532613.2 | 1 | 49 | XP_011530915.1 | |
| XM_011532614.2 | XP_011530916.1 | |||
| XM_011532615.2 | XP_011530917.2 | |||
| XM_011532616.2 | XP_011530918.1 | |||
| XM_011532617.2 | XP_011530919.1 | |||
| XM_011532618.2 | XP_011530920.1 | |||
| XM_011532619.2 | XP_011530921.1 | |||
| XM_017003474.1 | XP_016858963.1 | |||
| XM_017003475.1 | XP_016858964.1 | |||
| XM_017003476.1 | XP_016858965.1 | |||
| XM_017003477.1 | XP_016858966.1 | |||
| XM_017003478.1 | XP_016858967.1 | |||
| XM_017003479.1 | XP_016858968.1 | |||
| AF435956.1 | 1 | 48 | AAM20907.1 | |
| AK057516.1 | 1 | 50 | BAB71517.1 | |
| AK095462.1 | BAC04551.1 | |||
| AY358269.1 | AAQ88636.1 | |||
| BC024243.2 | AAH24243.3 | |||
| DB458482.1 | 1 | 59 | ||
| HY022533.1 | 1 | 69 |
Target Gene Details
Entrez Gene ID: | 101410544 |
Gene Name: | RMDN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:37950334-38036341 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN2-AS1 | NR_102712.1 | |||
| DB142860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6787n54 | Chr.2:37855471 - 37965002 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76027928] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map