Assay Details
Target Gene Details
Entrez Gene ID: | 83592 |
Gene Name: | aldo-keto reductase family 1 member E2 |
Gene Aliases: |
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR |
Location: |
Chr.10:4824554-4908421 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1E2 | NM_001040177.2 | NP_001035267.1 | ||
| NM_001271021.1 | NP_001257950.1 | |||
| NM_001271025.1 | NP_001257954.1 | |||
| NR_073125.1 | ||||
| NR_073126.1 | ||||
| NR_073127.1 | ||||
| XM_011519715.2 | XP_011518017.1 | |||
| XM_011519718.2 | XP_011518020.1 | |||
| XM_011519720.2 | XP_011518022.1 | |||
| XM_011519722.2 | XP_011518024.1 | |||
| XM_011519724.2 | XP_011518026.1 | |||
| XM_017016744.1 | XP_016872233.1 | |||
| XM_017016745.1 | XP_016872234.1 | |||
| AB040820.1 | BAC54565.1 | |||
| AB040821.1 | BAC54566.1 | |||
| AB040822.1 | BAC54567.1 | |||
| AB055603.1 | BAC54568.1 | |||
| AF263242.1 | AAK58523.1 | |||
| AK074211.1 | ||||
| BM724648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045155 | Chr.10:4830047 - 5169355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map