Assay Details
Target Gene Details
Entrez Gene ID: | 25896 |
Gene Name: | integrator complex subunit 7 |
Gene Aliases: |
C1orf73, INT7 |
Location: |
Chr.1:211940328-212035660 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS7 | NM_001199809.1 | 18 | 2582 | NP_001186738.1 |
| NM_001199811.1 | 19 | 2687 | NP_001186740.1 | |
| NM_001199812.1 | 19 | 2669 | NP_001186741.1 | |
| NM_015434.3 | 19 | 2729 | NP_056249.1 | |
| NR_037667.1 | 20 | 2763 | ||
| XM_011509396.2 | 20 | 2722 | XP_011507698.1 | |
| AK001363.1 | 19 | 2529 | BAA91650.1 | |
| AK001598.1 | 19 | 2611 | BAA91779.1 | |
| AK022509.1 | 19 | 2589 | BAB14067.1 | |
| AK022589.1 | 11 | 1396 | BAB14116.1 | |
| AK297225.1 | 18 | 2461 | ||
| AL117576.1 | 8 | 1003 | CAB56000.1 | |
| AL133049.2 | 14 | 1898 | CAB61376.1 | |
| BC020523.2 | 19 | 2563 | AAH20523.1 | |
| BC030716.1 | 19 | 2584 | AAH30716.1 | |
| BC033918.1 | 19 | 2588 | AAH33918.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76573676] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map