Assay Details
Target Gene Details
Entrez Gene ID: | 54888 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 2 |
Gene Aliases: |
MISU, MRT5, SAKI, TRM4 |
Location: |
Chr.5:6599239-6633360 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN2 | NM_001193455.1 | NP_001180384.1 | ||
| NM_017755.5 | NP_060225.4 | |||
| NR_037947.1 | ||||
| AK000310.1 | BAA91075.1 | |||
| AK023994.1 | BAB14762.1 | |||
| AK055456.1 | ||||
| AK291144.1 | ||||
| AK298980.1 | ||||
| BC001041.2 | AAH01041.3 | |||
| BC137083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894117 | Chr.5:6511201 - 6616164 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map