Assay Details
Target Gene Details
Entrez Gene ID: | 399694 |
Gene Name: | SHC adaptor protein 4 |
Gene Aliases: |
RaLP, SHCD |
Location: |
Chr.15:48823737-48963444 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHC4 | NM_203349.3 | 12 | 2433 | NP_976224.3 |
| XM_005254375.3 | 12 | 1590 | XP_005254432.1 | |
| AK124916.1 | 5 | 895 | ||
| AK125762.1 | 11 | 1394 | ||
| AK302156.1 | 10 | 1366 | ||
| AY358250.1 | 9 | 2106 | AAQ88617.1 | |
| BC033907.2 | 12 | 2433 | AAH33907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833001 | Chr.15:48751446 - 48912568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map