Assay Details
Target Gene Details
Entrez Gene ID: | 54972 |
Gene Name: | transmembrane protein 132A |
Gene Aliases: |
GBP, HSPA5BP1 |
Location: |
Chr.11:60924441-60937159 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM132A | NM_017870.3 | 11 | 3186 | NP_060340.2 |
| NM_178031.2 | 11 | 3183 | NP_821174.1 | |
| XM_017017951.1 | 10 | 4783 | XP_016873440.1 | |
| XM_017017952.1 | 10 | 4779 | XP_016873441.1 | |
| AK000546.1 | 9 | 2403 | BAA91245.1 | |
| AK023577.1 | 6 | 1902 | BAB14613.1 | |
| BC114569.1 | 11 | 3148 | AAI14570.1 |
More Information
Set Membership: |
|

Genomic Map