Assay Details
Target Gene Details
Entrez Gene ID: | 27076 |
Gene Name: | LY6/PLAUR domain containing 3 |
Gene Aliases: |
C4.4A |
Location: |
Chr.19:43460794-43465679 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYPD3 | NM_014400.2 | 3 | 395 | NP_055215.2 |
| AF082889.1 | 3 | 372 | AAD13751.1 | |
| AJ223603.2 | 3 | 323 | CAA11469.2 | |
| AK075464.1 | 3 | 324 | ||
| AK290905.1 | 3 | 324 | ||
| AK314773.1 | 3 | 324 | ||
| AL832153.1 | 3 | 325 | CAI46120.1 | |
| AY359006.1 | 3 | 302 | AAQ89365.1 | |
| BC039167.1 | 3 | 309 | AAH39167.1 | |
| BP367221.1 | 3 | 369 | ||
| DA837818.1 | 3 | 395 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2501 | Chr.19:43438429 - 43474272 on Build GRCh38 | Deletion |
|
| dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
|
| nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain |
|
| nsv509744 | Chr.19:43400292 - 43476817 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map