Assay Details
Target Gene Details
Entrez Gene ID: | 84240 |
Gene Name: | zinc finger CCHC-type containing 9 |
Gene Aliases: |
PPP1R41 |
Location: |
Chr.5:81301583-81313147 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZCCHC9 | NM_001131035.1 | NP_001124507.1 | ||
| NM_001131036.1 | NP_001124508.1 | |||
| NM_032280.2 | 1 | 114 | NP_115656.1 | |
| XM_005248616.3 | XP_005248673.1 | |||
| AK314160.1 | 1 | 56 | ||
| AL512712.1 | 1 | 94 | CAC21654.1 | |
| BC032736.1 | AAH32736.1 | |||
| BG776648.1 | ||||
| CB126982.1 | ||||
| CN261947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598750 | Chr.5:81207709 - 81303759 on Build GRCh38 | Gain |
|
| esv3890664 | Chr.5:81194463 - 81323227 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map