Assay Details
Target Gene Details
Entrez Gene ID: | 10542 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 5 |
Gene Aliases: |
HBXIP, XIP |
Location: |
Chr.1:110401253-110407924 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR5 | NM_006402.2 | NP_006393.2 | ||
| BC062619.1 | AAH62619.2 | |||
| BX092018.1 |
Target Gene Details
Entrez Gene ID: | 101410535 |
Gene Name: | LAMTOR5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:110407809-110416274 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR5-AS1 | NR_102697.1 | |||
| BE547292.1 | ||||
| BG718030.1 | ||||
| HY022359.1 |
More Information
Set Membership: |
|

Genomic Map