Assay Details
Target Gene Details
Entrez Gene ID: | 11170 |
Gene Name: | family with sequence similarity 107 member A |
Gene Aliases: |
DRR1, TU3A |
Location: |
Chr.3:58564112-58627610 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM107A | NM_001076778.2 | 4 | 2822 | NP_001070246.1 |
| NM_001282713.1 | 4 | 2622 | NP_001269642.1 | |
| NM_001282714.1 | 5 | 3070 | NP_001269643.1 | |
| NM_007177.3 | 5 | 2920 | NP_009108.1 | |
| AF089853.1 | 4 | 2913 | AAD16093.1 | |
| AF089854.1 | 5 | 3011 | AAD16094.1 | |
| BM980503.1 | 1 | 569 | ||
| EU569833.1 | 4 | 2477 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain |
|
| nsv470605 | Chr.3:58548399 - 58580265 on Build GRCh38 | Gain |
|
| nsv3839 | Chr.3:58519895 - 58564972 on Build GRCh38 | Deletion |
|
| esv3893702 | Chr.3:58557785 - 58858605 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map