Assay Details
Target Gene Details
Entrez Gene ID: | 230 |
Gene Name: | aldolase, fructose-bisphosphate C |
Gene Aliases: |
ALDC |
Location: |
Chr.17:28573115-28576963 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDOC | NM_005165.2 | NP_005156.1 | ||
| XM_005257949.2 | XP_005258006.1 | |||
| XM_011524556.1 | XP_011522858.1 | |||
| AF054987.1 | AAC09348.1 | |||
| AK293453.1 | ||||
| AK293623.1 | ||||
| AK293693.1 | ||||
| AK294091.1 | ||||
| AK294157.1 | ||||
| AK296039.1 | ||||
| AK296053.1 | ||||
| AK312281.1 | ||||
| BC003613.2 | ||||
| BC065565.1 | ||||
| BC103760.1 | AAI03761.1 | |||
| BC106925.1 | AAI06926.1 | |||
| BC106926.1 | AAI06927.1 | |||
| BT007006.1 | AAP35652.1 | |||
| CR541862.1 | CAG46660.1 | |||
| CR541881.1 | CAG46679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map