Assay Details
Target Gene Details
Entrez Gene ID: | 2936 |
Gene Name: | glutathione-disulfide reductase |
Gene Aliases: |
HEL-75, HEL-S-122m |
Location: |
Chr.8:30678061-30727969 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSR | NM_000637.3 | 5 | 677 | NP_000628.2 |
| NM_001195102.1 | 5 | 677 | NP_001182031.1 | |
| NM_001195103.1 | 5 | 677 | NP_001182032.1 | |
| NM_001195104.1 | 5 | 677 | NP_001182033.1 | |
| AB519179.1 | 5 | 543 | ||
| AB519180.1 | 5 | 543 | ||
| AB519181.1 | 5 | 543 | ||
| AF228704.1 | 5 | 543 | AAF37574.1 | |
| BC069244.1 | 5 | 634 | AAH69244.1 | |
| FJ224321.1 | 5 | 634 | ||
| GQ472231.1 | 5 | 634 | ||
| X15722.1 | 5 | 518 | CAA33744.1 | |
| X54507.1 | 3 | 177 | CAA38367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516544 | Chr.8:30678143 - 30703370 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map