Assay Details
Target Gene Details
Entrez Gene ID: | 516 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9) |
Gene Aliases: |
ATP5A, ATP5G |
Location: |
Chr.17:48892786-48895871 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5G1 | NM_001002027.1 | NP_001002027.1 | ||
| NM_005175.2 | NP_005166.1 | |||
| AK095669.1 | 2 | 1123 | ||
| AK311848.1 | ||||
| AL080089.1 | CAB45704.1 | |||
| BC004963.2 | AAH04963.1 | |||
| BF573360.1 | ||||
| BT007230.1 | AAP35894.1 | |||
| CF129854.1 | ||||
| CR533449.1 | CAG38480.1 | |||
| D13118.1 | BAA02420.1 |
Target Gene Details
Entrez Gene ID: | 105371814 |
Gene Name: | uncharacterized LOC105371814 |
Gene Aliases: |
- |
Location: |
Chr.17:48874860-48908983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371814 | NR_135674.1 | |||
| CX873301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map