Assay Details
Target Gene Details
Entrez Gene ID: | 7051 |
Gene Name: | transglutaminase 1 |
Gene Aliases: |
ARCI1, ICR2, KTG, LI, LI1, TGASE, TGK |
Location: |
Chr.14:24249114-24263210 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM1 | NM_000359.2 | NP_000350.1 | ||
| AK291350.1 | ||||
| AK299436.1 | ||||
| AK301652.1 | ||||
| AK315819.1 | ||||
| AK315828.1 | ||||
| AK315843.1 | ||||
| BC034699.1 | AAH34699.1 | |||
| D90287.1 | BAA14329.1 | |||
| M55183.1 | AAA59474.1 | |||
| M62925.1 | AAA61166.1 | |||
| X57974.1 | CAA41040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952818 | Chr.14:24249495 - 24270894 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map