Assay Details
Target Gene Details
Entrez Gene ID: | 196264 |
Gene Name: | myelin protein zero like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:118226690-118252368 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZL3 | NM_001286152.1 | 3 | 427 | NP_001273081.1 |
| NM_198275.2 | 3 | 463 | NP_938016.1 | |
| NR_104404.1 | ||||
| NR_104405.1 | ||||
| XM_006718775.2 | 3 | 815 | XP_006718838.1 | |
| XM_017017304.1 | 3 | 574 | XP_016872793.1 | |
| AK095399.1 | 3 | 415 | ||
| AK289390.1 | 3 | 415 | ||
| AK296951.1 | ||||
| AK304292.1 | 3 | 406 | ||
| AK310966.1 | ||||
| AY358626.1 | 3 | 391 | AAQ88989.1 | |
| BC113586.1 | 3 | 445 | ||
| BC143897.1 | 3 | 409 | ||
| DC417020.1 | 3 | 463 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511 | Chr.11:118204437 - 118239426 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map