Assay Details
Target Gene Details
Entrez Gene ID: | 10040 |
Gene Name: | target of myb1 like 1 membrane trafficking protein |
Gene Aliases: |
SRCASM |
Location: |
Chr.17:54900691-54961967 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOM1L1 | NM_001321173.1 | NP_001308102.1 | ||
| NM_001321174.1 | NP_001308103.1 | |||
| NM_001321175.1 | NP_001308104.1 | |||
| NM_001321176.1 | NP_001308105.1 | |||
| NM_005486.2 | NP_005477.2 | |||
| XM_017024002.1 | XP_016879491.1 | |||
| AJ010071.1 | CAA08993.1 | |||
| AK001893.1 | ||||
| AK223125.1 | BAD96845.1 | |||
| AK225351.1 | ||||
| AK300823.1 | ||||
| AK303912.1 | ||||
| AK303913.1 | ||||
| AK309164.1 | ||||
| AK315039.1 | ||||
| AK315907.1 | ||||
| BC029396.1 | AAH29396.1 | |||
| DC362418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3266n100 | Chr.17:54817463 - 55002289 on Build GRCh38 | Loss |
|
| esv3640828 | Chr.17:54782906 - 54915079 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map