Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIP1L1 | XM_005265769.4 | 19 | 3184 | XP_005265826.1 |
| XM_005265773.4 | 18 | 3117 | XP_005265830.1 | |
| XM_005265774.4 | 17 | 3029 | XP_005265831.1 | |
| XM_005265778.4 | 18 | 2955 | XP_005265835.1 | |
| XM_005265779.4 | 17 | 2883 | XP_005265836.1 | |
| XM_005265781.4 | 17 | 3001 | XP_005265838.1 | |
| XM_005265782.4 | 16 | 2920 | XP_005265839.1 | |
| XM_017008662.1 | 17 | 3062 | XP_016864151.1 | |
| XM_017008663.1 | 18 | 3018 | XP_016864152.1 | |
| XM_017008664.1 | 17 | 3073 | XP_016864153.1 | |
| XM_017008665.1 | 17 | 2989 | XP_016864154.1 | |
| XM_017008666.1 | 16 | 3006 | XP_016864155.1 | |
| XM_017008667.1 | 17 | 2972 | XP_016864156.1 | |
| XM_017008668.1 | 16 | 3018 | XP_016864157.1 | |
| XM_017008669.1 | 16 | 2918 | XP_016864158.1 | |
| XM_017008670.1 | 16 | 2814 | XP_016864159.1 | |
| XM_017008671.1 | 16 | 2733 | XP_016864160.1 | |
| AK090938.1 | 7 | 1884 |
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | NM_001126328.2 | 11 | 2811 | NP_001119800.1 |
| NM_032622.2 | 10 | 2542 | NP_116011.2 | |
| XM_005265785.4 | 11 | 2969 | XP_005265842.1 | |
| XM_017008776.1 | 12 | 3030 | XP_016864265.1 | |
| AF237782.1 | 10 | 2473 | AAK15039.1 | |
| AY358326.1 | 10 | 2453 | AAQ88692.1 | |
| BC022983.1 | 11 | 2687 | AAH22983.1 | |
| BC034737.1 | 10 | 2462 | AAH34737.1 |
More Information
Set Membership: |
|

Genomic Map