Assay Details
Target Gene Details
Entrez Gene ID: | 283551 |
Gene Name: | long intergenic non-protein coding RNA 1588 |
Gene Aliases: |
C14orf182 |
Location: |
Chr.14:49981669-50007520 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01588 | NR_126496.1 | 1 | 60 | |
| NR_126497.1 | 1 | 204 | ||
| NR_126498.1 | 1 | 1061 | ||
| NR_126500.1 | 1 | 204 | ||
| AK090420.1 | 1 | 60 | BAC03401.1 | |
| AK091232.1 | 1 | 1061 | ||
| BC128083.1 | 1 | 784 | ||
| BC144351.1 | 1 | 204 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map