Assay Details
Target Gene Details
Entrez Gene ID: | 84820 |
Gene Name: | RNA polymerase II subunit J4, pseudogene |
Gene Aliases: |
RPB11-phi |
Location: |
Chr.7:43940895-44019194 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2J4 | NR_003655.3 | 16 | 4564 | |
| BX537878.1 | 16 | 4519 |
Target Gene Details
Entrez Gene ID: | 51619 |
Gene Name: | ubiquitin conjugating enzyme E2 D4 (putative) |
Gene Aliases: |
HBUCE1 |
Location: |
Chr.7:43926436-43956136 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2D4 | NM_015983.3 | NP_057067.1 | ||
| XM_006715734.3 | XP_006715797.1 | |||
| XM_011515420.2 | XP_011513722.1 | |||
| XM_011515422.2 | XP_011513724.1 | |||
| XM_011515423.2 | XP_011513725.1 | |||
| XM_017012292.1 | XP_016867781.1 | |||
| XM_017012293.1 | XP_016867782.1 | |||
| XM_017012294.1 | XP_016867783.1 | |||
| XM_017012295.1 | XP_016867784.1 | |||
| AF125044.1 | AAD31180.1 | |||
| AF125045.1 | AAD31181.1 | |||
| AK001446.1 | BAA91697.1 | |||
| BC004104.2 | AAH04104.1 | |||
| BG499454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759527 | Chr.7:43912940 - 44017192 on Build GRCh38 | Gain |
|
| nsv1032143 | Chr.7:43931542 - 43948877 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv1016724 | Chr.7:43927631 - 43957476 on Build GRCh38 | Gain |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map