Assay Details
Target Gene Details
Entrez Gene ID: | 55897 |
Gene Name: | mesoderm posterior bHLH transcription factor 1 |
Gene Aliases: |
bHLHc5 |
Location: |
Chr.15:89734772-89751309 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESP1 | NM_018670.3 | 2 | 897 | NP_061140.1 |
| AL357533.1 | 2 | 450 | CAB93425.1 | |
| AL357534.1 | 2 | 428 | CAB93426.1 | |
| AL357535.1 | CAB93427.1 | |||
| AL357570.1 | 2 | 344 | ||
| AL357571.1 | 2 | 717 | ||
| BC006219.1 | 2 | 905 | AAH06219.1 | |
| BP212916.1 | 2 | 467 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952025 | Chr.15:89742870 - 89762169 on Build GRCh38 | Deletion |
|
| nsv570388 | Chr.15:89730911 - 89794962 on Build GRCh38 | Loss |
|
| nsv1046117 | Chr.15:89683716 - 89765410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map