Assay Details
Target Gene Details
Entrez Gene ID: | 81544 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 5 |
Gene Aliases: |
GDE2, PP1665 |
Location: |
Chr.11:75434640-75525916 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDPD5 | NM_030792.6 | 17 | 3120 | NP_110419.5 |
| XM_006718697.3 | 18 | 3726 | XP_006718760.1 | |
| XM_011545276.2 | 19 | 3515 | XP_011543578.1 | |
| XM_011545277.2 | 19 | 3903 | XP_011543579.1 | |
| XM_011545278.2 | 19 | 3966 | XP_011543580.1 | |
| XM_011545279.2 | 17 | 3485 | XP_011543581.1 | |
| XM_011545280.1 | 16 | 2818 | XP_011543582.1 | |
| XM_011545281.2 | 12 | 2638 | XP_011543583.1 | |
| XM_011545285.2 | 13 | 4574 | XP_011543587.1 | |
| XM_011545286.1 | 12 | 2374 | XP_011543588.1 | |
| XM_011545287.1 | 12 | 2371 | XP_011543589.1 | |
| AF217989.1 | 5 | 1455 | AAG17232.1 | |
| AF289614.1 | 1 | 561 | AAL55798.1 | |
| AF318351.1 | 17 | 2976 | AAL55858.1 | |
| AF318377.1 | 17 | 3389 | AAL55884.1 | |
| AF451987.1 | 10 | 2134 | AAP97686.1 | |
| AK074848.1 | 11 | 2208 | BAC11242.1 | |
| AK098808.1 | 12 | 2371 | BAC05418.1 | |
| AL833941.1 | 18 | 3157 | CAD38796.2 | |
| AY358477.1 | 12 | 2387 | AAQ88841.1 | |
| BC018771.1 | 18 | 3168 | AAH18771.1 | |
| BC020203.1 | 1 | 975 | AAH20203.1 | |
| BC029416.1 | 1 | 731 | AAH29416.1 | |
| BC033391.1 | 13 | 4110 | AAH33391.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79131751] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map