Assay Details
Target Gene Details
Entrez Gene ID: | 271 |
Gene Name: | adenosine monophosphate deaminase 2 |
Gene Aliases: |
PCH9, SPG63 |
Location: |
Chr.1:109619813-109632055 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMPD2 | NM_001257360.1 | 1 | 152 | NP_001244289.1 |
| NM_139156.3 | 1 | 152 | NP_631895.1 | |
| AK025706.1 | 1 | 128 | ||
| AK308907.1 | ||||
| AK314494.1 | 1 | 75 | ||
| BC007711.2 | 1 | 77 | AAH07711.1 | |
| BC075844.1 | 1 | 77 | AAH75844.1 | |
| DA558264.1 | 1 | 152 | ||
| U16267.1 | 1 | 72 | AAC50306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2299 | Chr.1:109610296 - 109662341 on Build GRCh38 | Deletion |
|
| dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map