Assay Details
Target Gene Details
Entrez Gene ID: | 2674 |
Gene Name: | GDNF family receptor alpha 1 |
Gene Aliases: |
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1 |
Location: |
Chr.10:116056925-116273645 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA1 | NM_001145453.2 | 10 | 2696 | NP_001138925.1 |
| NM_005264.5 | 11 | 2711 | NP_005255.1 | |
| NM_145793.3 | 9 | 2245 | NP_665736.1 | |
| XM_005269696.1 | 11 | 2321 | XP_005269753.1 | |
| XM_011539634.1 | 11 | 2286 | XP_011537936.1 | |
| AF038421.1 | 12 | 2501 | AAC39692.1 | |
| BM675677.1 | 1 | 390 | ||
| U95847.1 | AAB71811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1995539 | Chr.10:116063839 - 116064251 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map