Assay Details
Target Gene Details
Entrez Gene ID: | 55697 |
Gene Name: | Vac14, PIKFYVE complex component |
Gene Aliases: |
ArPIKfyve, TAX1BP2, TRX |
Location: |
Chr.16:70687439-70801169 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAC14 | NM_018052.3 | NP_060522.3 | ||
| XM_005256038.3 | XP_005256095.1 | |||
| XM_005256041.3 | XP_005256098.2 | |||
| XM_011523223.2 | XP_011521525.1 | |||
| XM_011523224.2 | XP_011521526.1 | |||
| XM_011523225.2 | XP_011521527.1 | |||
| XM_017023441.1 | XP_016878930.1 | |||
| XM_017023442.1 | XP_016878931.1 | |||
| AK025479.1 | BAB15145.1 | |||
| AK056433.1 | ||||
| AK297564.1 | ||||
| AK307766.1 | ||||
| BC007214.1 | AAH07214.2 | |||
| BC125108.1 | ||||
| BC125109.1 | ||||
| CR457343.1 | CAG33624.1 |
Target Gene Details
Entrez Gene ID: | 100130894 |
Gene Name: | VAC14 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:70755060-70773251 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAC14-AS1 | NR_034083.2 | 6 | 1130 | |
| AK095020.1 | 6 | 1056 | BAC04476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
| nsv517391 | Chr.16:70632507 - 70806587 on Build GRCh38 | Loss |
|
| nsv572945 | Chr.16:70632819 - 70806587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map