Assay Details
Target Gene Details
Entrez Gene ID: | 23237 |
Gene Name: | activity regulated cytoskeleton associated protein |
Gene Aliases: |
Arg3.1 |
Location: |
Chr.8:142611044-142614472 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARC | NM_015193.4 | 1 | 1169 | NP_056008.1 |
| AF193421.1 | 1 | 1157 | AAF07185.1 | |
| BC012321.1 | 1 | 1173 | AAH12321.1 | |
| BC075802.1 | 1 | 1161 | AAH75802.1 | |
| D87468.1 | 1 | 1161 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470245 | Chr.8:142499525 - 142634680 on Build GRCh38 | Loss |
|
| nsv950002 | Chr.8:142055440 - 142642221 on Build GRCh38 | Deletion |
|
| esv2661137 | Chr.8:142256294 - 142935448 on Build GRCh38 | Deletion |
|
| nsv612709 | Chr.8:142564638 - 142670446 on Build GRCh38 | Loss |
|
| dgv7335n100 | Chr.8:142430053 - 142616194 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map