Assay Details
Target Gene Details
Entrez Gene ID: | 55554 |
Gene Name: | kallikrein related peptidase 15 |
Gene Aliases: |
ACO, HSRNASPH |
Location: |
Chr.19:50825289-50834337 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK15 | NM_001277081.1 | 5 | 720 | NP_001264010.1 |
| NM_001277082.1 | 4 | 583 | NP_001264011.1 | |
| NM_017509.3 | 5 | 723 | NP_059979.2 | |
| NR_102274.1 | 5 | 602 | ||
| XM_006723265.3 | 6 | 1524 | XP_006723328.1 | |
| XM_011527085.2 | 6 | 1520 | XP_011525387.1 | |
| XM_011527087.2 | 5 | 1273 | XP_011525389.1 | |
| XM_011527088.2 | XP_011525390.1 | |||
| XM_011527089.2 | 5 | 1394 | XP_011525391.1 | |
| AF303046.1 | 5 | 722 | AAK62813.1 | |
| AY373373.1 | 4 | 812 | AAQ82620.1 | |
| AY373374.1 | 4 | 675 | AAQ82621.1 | |
| BC069480.1 | 5 | 720 | AAH69480.1 | |
| BC069507.1 | 5 | 723 | AAH69507.1 | |
| BC069518.1 | 5 | 720 | AAH69518.1 | |
| BC126137.1 | 5 | 713 | ||
| BC144046.1 | 5 | 710 | ||
| X75363.1 | 1 | 1278 | CAA53145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv204n21 | Chr.19:50817827 - 50832941 on Build GRCh38 | Loss |
|
| esv3583464 | Chr.19:50818135 - 50842312 on Build GRCh38 | Loss |
|
| dgv203n21 | Chr.19:50797973 - 50832687 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map