Assay Details
Target Gene Details
Entrez Gene ID: | 56919 |
Gene Name: | DEAH-box helicase 33 |
Gene Aliases: |
DDX33 |
Location: |
Chr.17:5440912-5469060 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX33 | NM_001199699.1 | 11 | 3416 | NP_001186628.1 |
| NM_020162.3 | 12 | 3577 | NP_064547.2 | |
| XM_017024877.1 | 8 | 2453 | XP_016880366.1 | |
| AK026944.1 | 6 | 2094 | BAB15596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map