Assay Details
Target Gene Details
Entrez Gene ID: | 9682 |
Gene Name: | lysine demethylase 4A |
Gene Aliases: |
JHDM3A, JMJD2, JMJD2A, TDRD14A |
Location: |
Chr.1:43650118-43705518 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4A | NM_014663.2 | 2 | 171 | NP_055478.2 |
| XM_005271354.3 | 2 | 208 | XP_005271411.1 | |
| XM_005271355.3 | 2 | 126 | XP_005271412.1 | |
| XM_005271356.3 | 2 | 110 | XP_005271413.1 | |
| XM_017002912.1 | 2 | 179 | XP_016858401.1 | |
| AB014577.1 | 2 | 130 | ||
| AK300039.1 | 2 | 163 | ||
| BC002558.2 | 2 | 139 | AAH02558.1 | |
| BP198064.1 | 2 | 171 |
More Information
Set Membership: |
|

Genomic Map