Assay Details
Target Gene Details
Entrez Gene ID: | 11226 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 6 |
Gene Aliases: |
GALNAC-T6, GalNAcT6 |
Location: |
Chr.12:51351870-51391955 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT6 | NM_007210.3 | 8 | 1634 | NP_009141.2 |
| XM_005268607.1 | 8 | 1893 | XP_005268664.1 | |
| XM_006719214.2 | 7 | 1468 | XP_006719277.1 | |
| XM_011537819.2 | 9 | 1849 | XP_011536121.1 | |
| XM_011537822.2 | 8 | 1570 | XP_011536124.1 | |
| XM_017018744.1 | 8 | 1707 | XP_016874233.1 | |
| XM_017018745.1 | 9 | 1692 | XP_016874234.1 | |
| AB265820.1 | 7 | 1519 | ||
| AK025961.1 | BAB15297.1 | |||
| AK074658.1 | 7 | 1532 | BAC11118.1 | |
| AK092496.1 | 1 | 1040 | ||
| AK296852.1 | 9 | 1540 | ||
| BC035822.1 | 8 | 1637 | AAH35822.2 | |
| BC114505.1 | 6 | 1313 | AAI14506.1 | |
| Y08565.1 | 6 | 1313 | CAA69876.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832408 | Chr.12:51302821 - 51477555 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map