Assay Details
Target Gene Details
Entrez Gene ID: | 128229 |
Gene Name: | TSSK6 activating cochaperone |
Gene Aliases: |
C1orf182, SIP, SSTK-IP |
Location: |
Chr.1:156337314-156346995 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSACC | NM_001304817.1 | NP_001291746.1 | ||
| NM_001304818.1 | NP_001291747.1 | |||
| NM_001304819.1 | NP_001291748.1 | |||
| NM_001304820.1 | NP_001291749.1 | |||
| NM_001304821.1 | NP_001291750.1 | |||
| NM_001304822.1 | NP_001291751.1 | |||
| NM_001304823.1 | NP_001291752.1 | |||
| NM_001304824.1 | NP_001291753.1 | |||
| NM_001304825.1 | NP_001291754.1 | |||
| NM_001304826.1 | NP_001291755.1 | |||
| NM_144627.4 | NP_653228.1 | |||
| NR_130913.1 | ||||
| AI336861.1 | ||||
| AK311950.1 | ||||
| AY048672.1 | AAL07513.1 | |||
| AY048673.1 | AAL07514.1 | |||
| BC014605.1 | AAH14605.1 | |||
| BE464129.1 | ||||
| BI831108.1 | ||||
| DB446801.1 | ||||
| DQ577676.1 | ||||
| HY019225.1 | ||||
| HY034095.1 | ||||
| HY044771.1 | ||||
| HY048925.1 | ||||
| HY230397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587635 | Chr.1:156331944 - 156348237 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map