Assay Details
Target Gene Details
Entrez Gene ID: | 92154 |
Gene Name: | metastasis suppressor 1 like |
Gene Aliases: |
ABBA-1, ABBA1 |
Location: |
Chr.16:70661204-70686207 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTSS1L | NM_138383.2 | NP_612392.1 | ||
| XM_005256236.4 | XP_005256293.2 | |||
| XM_011523450.2 | XP_011521752.1 | |||
| XM_011523453.2 | XP_011521755.2 | |||
| XM_011523454.2 | XP_011521756.2 | |||
| XM_017023879.1 | XP_016879368.1 | |||
| XM_017023880.1 | XP_016879369.1 | |||
| XM_017023881.1 | XP_016879370.1 | |||
| XM_017023882.1 | XP_016879371.1 | |||
| XM_017023883.1 | XP_016879372.1 | |||
| XM_017023884.1 | XP_016879373.1 | |||
| AB115770.1 | BAC98378.1 | |||
| BC002770.1 | AAH02770.1 | |||
| BU859607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952049 | Chr.16:70674798 - 70680097 on Build GRCh38 | Deletion |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
| nsv517391 | Chr.16:70632507 - 70806587 on Build GRCh38 | Loss |
|
| nsv1856 | Chr.16:70670900 - 70684358 on Build GRCh38 | Insertion |
|
| nsv572945 | Chr.16:70632819 - 70806587 on Build GRCh38 | Gain |
|
| nsv1059226 | Chr.16:70674795 - 70745241 on Build GRCh38 | Gain |
|
| nsv471094 | Chr.16:70660097 - 70716067 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map