Assay Details
Target Gene Details
Entrez Gene ID: | 84138 |
Gene Name: | solute carrier family 7 member 6 opposite strand |
Gene Aliases: |
- |
Location: |
Chr.16:68300615-68310965 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6OS | NM_032178.2 | NP_115554.2 | ||
| XM_011523372.2 | 4 | 791 | XP_011521674.1 | |
| AK023353.1 | BAB14538.1 | |||
| AK091674.1 | ||||
| AK225712.1 | ||||
| AY078070.1 | AAL79958.1 | |||
| BC013778.1 | AAH13778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map