Assay Details
Target Gene Details
Entrez Gene ID: | 4150 |
Gene Name: | MYC associated zinc finger protein |
Gene Aliases: |
PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87 |
Location: |
Chr.16:29806096-29811183 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAZ | NM_001042539.2 | 6 | 2680 | NP_001036004.1 |
| NM_001276275.1 | 6 | 2270 | NP_001263204.1 | |
| NM_001276276.1 | 3 | 1540 | NP_001263205.1 | |
| NM_002383.3 | 5 | 2455 | NP_002374.2 | |
| NR_074080.1 | 4 | 1604 | ||
| XM_006721047.3 | 7 | 2467 | XP_006721110.1 | |
| AA588396.1 | 1 | 95 | ||
| AB209061.1 | 1 | 3598 | BAD92298.1 | |
| AI079560.1 | 1 | 82 | ||
| AI360548.1 | 1 | 105 | ||
| AK074720.1 | 3 | 1306 | ||
| BC041629.1 | 5 | 1830 | AAH41629.1 | |
| BM695402.1 | 1 | 362 | ||
| BQ431307.1 | 1 | 157 | ||
| BU615653.1 | 1 | 109 | ||
| BX363391.2 | 1 | 754 | ||
| FJ532357.1 | 6 | 2260 | ||
| M94046.1 | 5 | 2312 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833186 | Chr.16:29719552 - 29863674 on Build GRCh38 | Loss |
|
| nsv953800 | Chr.16:29804780 - 29816179 on Build GRCh38 | Deletion |
|
| nsv827611 | Chr.16:29808186 - 29830422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map