Assay Details
Target Gene Details
Entrez Gene ID: | 5507 |
Gene Name: | protein phosphatase 1 regulatory subunit 3C |
Gene Aliases: |
PPP1R5 |
Location: |
Chr.10:91628440-91633101 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3C | NM_005398.5 | 2 | 2414 | NP_005389.1 |
| AI766011.1 | 1 | 163 | ||
| BC012625.1 | 2 | 2339 | AAH12625.1 | |
| BX537399.1 | 2 | 2415 | CAD97641.1 |
More Information
Set Membership: |
|

Genomic Map