Assay Details
Target Gene Details
Entrez Gene ID: | 6812 |
Gene Name: | syntaxin binding protein 1 |
Gene Aliases: |
MUNC18-1, NSEC1, P67, RBSEC1, UNC18 |
Location: |
Chr.9:127612207-127692716 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STXBP1 | NM_001032221.3 | NP_001027392.1 | ||
| NM_003165.3 | NP_003156.1 | |||
| AF004562.1 | AAC39688.1 | |||
| AF004563.1 | AAC39689.1 | |||
| AK293984.1 | ||||
| AK295154.1 | ||||
| AK295802.1 | ||||
| BC015749.1 | AAH15749.1 | |||
| D63851.1 | BAA19483.1 | |||
| DC325017.1 | ||||
| Y12723.1 | CAA73255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1075675 | Chr.9:127611820 - 127613021 on Build GRCh38 | Deletion |
|
| nsv951767 | Chr.9:127612122 - 127613121 on Build GRCh38 | Deletion |
|
| nsv1126921 | Chr.9:127612321 - 127613021 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map