Assay Details
Target Gene Details
Entrez Gene ID: | 54438 |
Gene Name: | glucose-fructose oxidoreductase domain containing 1 |
Gene Aliases: |
ADG-90, C6orf114 |
Location: |
Chr.6:13357830-13487637 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFOD1 | NM_001242628.1 | 2 | 2195 | NP_001229557.1 |
| NM_001242630.1 | 2 | 2250 | NP_001229559.1 | |
| NM_018988.3 | 2 | 3022 | NP_061861.1 | |
| XM_011514699.2 | 2 | 2211 | XP_011513001.1 | |
| AK000337.1 | 2 | 2940 | BAA91093.1 | |
| AK027010.1 | 2 | 2250 | BAB15624.1 | |
| AW137218.1 | 1 | 304 |
More Information
Set Membership: |
|

Genomic Map