Assay Details
Target Gene Details
Entrez Gene ID: | 57591 |
Gene Name: | megakaryoblastic leukemia (translocation) 1 |
Gene Aliases: |
BSAC, MAL, MRTF-A |
Location: |
Chr.22:40410281-40636719 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKL1 | NM_001282660.1 | NP_001269589.1 | ||
| NM_001282661.1 | NP_001269590.1 | |||
| NM_001282662.1 | NP_001269591.1 | |||
| NM_001318139.1 | NP_001305068.1 | |||
| NM_020831.4 | NP_065882.1 | |||
| XM_005261692.1 | XP_005261749.1 | |||
| XM_005261694.1 | XP_005261751.1 | |||
| XM_011530283.1 | XP_011528585.1 | |||
| XM_011530285.1 | XP_011528587.1 | |||
| XM_011530287.1 | XP_011528589.1 | |||
| XM_017028888.1 | XP_016884377.1 | |||
| XM_017028889.1 | XP_016884378.1 | |||
| AB037859.2 | ||||
| AF368061.1 | AAK54721.1 | |||
| AJ297257.1 | CAC38826.1 | |||
| AJ297258.1 | CAC38827.1 | |||
| AL713710.1 | CAD28507.2 | |||
| BC114364.2 | AAI14365.1 | |||
| BC115039.1 | ||||
| CR456522.1 | CAG30408.1 | |||
| HG764171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829233 | Chr.22:40404227 - 40438253 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map