Assay Details
Target Gene Details
Entrez Gene ID: | 80347 |
Gene Name: | Coenzyme A synthase |
Gene Aliases: |
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2 |
Location: |
Chr.17:42562074-42566281 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COASY | NM_001042529.2 | 5 | 1320 | NP_001035994.1 |
| NM_001042532.3 | 6 | 1452 | NP_001035997.2 | |
| NM_025233.6 | 4 | 1712 | NP_079509.5 | |
| XM_006722116.3 | 5 | 1346 | XP_006722179.1 | |
| XM_011525300.1 | 5 | 1516 | XP_011523602.1 | |
| XM_017025167.1 | XP_016880656.1 | |||
| XM_017025168.1 | 5 | 925 | XP_016880657.1 | |
| AF208536.1 | 4 | 672 | AAF87955.1 | |
| AF453478.1 | 4 | 1163 | AAL50813.1 | |
| AK075415.1 | 4 | 1620 | ||
| AK092498.1 | 2 | 1387 | ||
| AK225485.1 | 5 | 1249 | ||
| AK297153.1 | 6 | 1375 | ||
| AK303766.1 | ||||
| AK314076.1 | 6 | 1376 | ||
| AL360167.1 | 4 | 672 | ||
| AL360168.1 | ||||
| AL833588.1 | 5 | 1244 | ||
| AY094602.1 | 5 | 1273 | AAM19996.1 | |
| BC006354.1 | 3 | 413 | AAH06354.1 | |
| BC020985.1 | 4 | 888 | AAH20985.1 | |
| BC067254.1 | 4 | 1616 | AAH67254.1 | |
| BT007168.1 | 2 | 146 | AAP35832.1 | |
| U18919.1 | 2 | 172 | AAA69699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map