Assay Details
Target Gene Details
Entrez Gene ID: | 5274 |
Gene Name: | serpin family I member 1 |
Gene Aliases: |
PI12, neuroserpin |
Location: |
Chr.3:167735335-167825569 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINI1 | NM_001122752.1 | 1 | 117 | NP_001116224.1 |
| NM_005025.4 | NP_005016.1 | |||
| XM_017006618.1 | 1 | 426 | XP_016862107.1 | |
| AK290082.1 | ||||
| BC018043.1 | AAH18043.1 | |||
| CR627434.1 | CAH10520.1 | |||
| DA270615.1 | ||||
| DA359846.1 | 1 | 117 | ||
| Z81326.1 | CAB03626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4105 | Chr.3:167723508 - 167754073 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map