Assay Details
Target Gene Details
Entrez Gene ID: | 51455 |
Gene Name: | REV1, DNA directed polymerase |
Gene Aliases: |
AIBP80, REV1L |
Location: |
Chr.2:99400475-99490123 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REV1 | NM_001037872.2 | 18 | 3132 | NP_001032961.1 |
| NM_001321454.1 | 19 | 3243 | NP_001308383.1 | |
| NM_001321455.1 | 19 | 3289 | NP_001308384.1 | |
| NM_001321458.1 | 19 | 3259 | NP_001308387.1 | |
| NM_001321459.1 | 19 | 3335 | NP_001308388.1 | |
| NM_001321460.1 | 19 | 3332 | NP_001308389.1 | |
| NM_016316.3 | 18 | 3135 | NP_057400.1 | |
| NR_135649.1 | 19 | 3332 | ||
| NR_135650.1 | 19 | 3289 | ||
| NR_135651.1 | 17 | 2966 | ||
| NR_135652.1 | 18 | 3135 | ||
| NR_135653.1 | 17 | 2969 | ||
| XM_011511341.2 | 12 | 1752 | XP_011509643.1 | |
| XM_017004309.1 | 19 | 3656 | XP_016859798.1 | |
| XM_017004310.1 | 19 | 3022 | XP_016859799.1 | |
| XM_017004311.1 | 19 | 3022 | XP_016859800.1 | |
| XM_017004312.1 | 19 | 3235 | XP_016859801.1 | |
| XM_017004313.1 | 18 | 4861 | XP_016859802.1 | |
| XM_017004314.1 | 15 | 2512 | XP_016859803.1 | |
| XM_017004315.1 | 15 | 2496 | XP_016859804.1 | |
| AB047646.1 | 18 | 3057 | BAB21441.1 | |
| AF151538.1 | 18 | 3088 | AAF06731.1 | |
| AF206019.1 | 18 | 3097 | AAF18986.1 | |
| AF357886.1 | 18 | 3053 | AAK43708.1 | |
| AJ131720.1 | 10 | 1454 | CAB38231.1 | |
| AK025176.1 | 10 | 1414 | ||
| AK307719.1 | 18 | 3046 | ||
| BC037734.1 | 19 | 3270 | AAH37734.1 | |
| BC130411.1 | 17 | 2903 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524466 | Chr.2:99335623 - 99415089 on Build GRCh38 | Loss |
|
| dgv4032n100 | Chr.2:99364388 - 99481422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map