Assay Details
Target Gene Details
Entrez Gene ID: | 158248 |
Gene Name: | tetratricopeptide repeat domain 16 |
Gene Aliases: |
- |
Location: |
Chr.9:127716064-127731600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|

Genomic Map