Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF1 | NM_001278682.1 | NP_001265611.1 | ||
| NM_001278684.1 | NP_001265613.1 | |||
| NM_001278686.1 | NP_001265615.1 | |||
| NM_003244.3 | NP_003235.1 | |||
| NM_173207.2 | NP_775299.1 | |||
| NM_173208.2 | NP_775300.1 | |||
| NM_173209.2 | NP_775301.1 | |||
| NM_174886.2 | NP_777480.1 | |||
| XM_017025958.1 | XP_016881447.1 | |||
| XM_017025959.1 | XP_016881448.1 | |||
| AF179900.1 | AAF81643.1 | |||
| AI813434.1 | ||||
| AK291112.1 | ||||
| AK309471.1 | ||||
| AU310737.1 | ||||
| BC000814.1 | AAH00814.1 | |||
| BE296707.1 | ||||
| BE906361.1 | ||||
| BF446044.1 | ||||
| BM849453.1 | ||||
| BQ437864.1 | ||||
| BU657748.1 | ||||
| CB161219.1 | ||||
| CB996365.1 | ||||
| CR450333.1 | CAG29329.1 | |||
| DR007147.1 | ||||
| HY041619.1 | ||||
| X89750.1 | CAA61897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527676 | Chr.18:3445828 - 3452980 on Build GRCh38 | Loss |
|
| nsv516490 | Chr.18:3448764 - 3451764 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map